missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. C049825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C049825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C049825G Supplier TCI America Supplier No. C049825G

Chemical Identifiers

CAS 6531-86-8
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00021278
InChI Key YTWWBKKPWACPPB-UHFFFAOYSA-N
PubChem CID 94327
IUPAC Name 2-cyclohexylcyclohexan-1-ol
SMILES C1CCC(CC1)C2CCCCC2O

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 g
Formula Weight 182.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexylcyclohexanol (cis- and trans- mixture)