missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylaminoethanesulfonic Acid 99.0+%, TCI America™

Catalog No. C092025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C092025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C092025G Supplier TCI America Supplier No. C092025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
Synonym n-cyclohexyltaurine, ches, 2-cyclohexylamino ethanesulfonic acid, 2-n-cyclohexylamino ethanesulfonic acid, ethanesulfonic acid, 2-cyclohexylamino, unii-71x53v3rz1, taurine, n-cyclohexyl, 2-cyclohexylamino ethane sulphonic acid, cyclohexylaminoethane sulfonic acid, 2-cyclohexylamino ethanesulfonicacid
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 207.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyclohexylaminoethanesulfonic Acid [Good′s buffer component for biological research]