missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-one, 97%

Catalog No. AAA1543514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543514 25 g
AAA1543522 100 g
AAA1543536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1543514 Supplier Thermo Scientific Chemicals Supplier No. A1543514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 930-68-7 | C6H8O | 96.13 g/mol

2-Cyclohexen-1-one is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclohexen-1-one is used as intermediates.

Solubility
Soluble in water, alcohols, benzene and acetone.

Notes
Store away from oxidizing agents and strong bases. Incompatible with reducing agents and bases.

Chemical Identifiers

CAS 930-68-7
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00001577
InChI Key FWFSEYBSWVRWGL-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one
PubChem CID 13594
ChEBI CHEBI:15977
IUPAC Name cyclohex-2-en-1-one
SMILES O=C1CCCC=C1

Specifications

Melting Point -53°C
Density 0.993
Boiling Point 167°C to 169°C
Flash Point 56°C (132°F)
Refractive Index 1.488
Quantity 25 g
UN Number UN2929
Beilstein 1280477
Solubility Information Soluble in water,alcohols,benzene and acetone.
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 2-Cyclohexen-1-one
Show More Show Less
Hazard Category H226-H301-H310+H330-H319
Hazard Statement GHS H Statement
H301-H310-H330-H226-H319
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 213-223-5
RTECSNumber GW7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only