missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyridine, 99%

Catalog No. aaa1290036
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Catalog No. Quantity
AAA1290036 500 g
AAA1290022 100 g
2 options

Catalog No. AAA1290036

Supplier: Thermo Scientific Chemicals A1290036

Only null left

CAS: 100-70-9 | C6H4N2 | 104.112 g/mol

It is employed as an important chemical intermediate for rimiterol hydrobromide and used as a bronchodilator. It is also used as intermediates of pharmaceutical and dye and pigment. It acts as a precursor of the respective amidate for protein modification via amidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an important chemical intermediate for rimiterol hydrobromide and used as a bronchodilator. It is also used as intermediates of pharmaceutical and dye and pigment. It acts as a precursor of the respective amidate for protein modification via amidation.

Solubility
Soluble in water at 20°C 67g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 100-70-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.112
MDL Number MFCD00006218
InChI Key FFNVQNRYTPFDDP-UHFFFAOYSA-N
Synonym 2-cyanopyridine, picolinonitrile, 2-pyridinecarbonitrile, pyridinecarbonitrile, 2-pyridyl nitrile, picolinic acid nitrile, cyanopyridine, unii-whr1dpg7ys, 2-pyridinecarboxylic acid, nitrile, 2-cyano pyridine
PubChem CID 7522
ChEBI CHEBI:27837
IUPAC Name pyridine-2-carbonitrile
SMILES C1=CC=NC(=C1)C#N

Specifications

Melting Point 26°C to 28°C
Density 1.081
pH 8.4
Boiling Point 212°C to 214°C
Flash Point 89°C (192°F)
Odor Characteristic
Refractive Index 1.529
Quantity 500 g
Beilstein 107710
Solubility Information Soluble in water at 20°C 67g/L.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material 2-Cyanopyridine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 202-880-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only