missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyridine, 99%

Catalog No. p-95059
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110850050 5 g Glass bottle
AC110851000 100 g Glass bottle
AC110855000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110850050 Supplier Thermo Scientific Chemicals Supplier No. 110850050
Only null left

CAS: 100-70-9 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-70-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006218
InChI Key FFNVQNRYTPFDDP-UHFFFAOYSA-N
Synonym 2-cyanopyridine, picolinonitrile, 2-pyridinecarbonitrile, pyridinecarbonitrile, 2-pyridyl nitrile, picolinic acid nitrile, cyanopyridine, unii-whr1dpg7ys, 2-pyridinecarboxylic acid, nitrile, 2-cyano pyridine
PubChem CID 7522
ChEBI CHEBI:27837
IUPAC Name pyridine-2-carbonitrile
SMILES C1=CC=NC(=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 26.0°C to 28.0°C
Color White to Brown
Boiling Point 212.0°C to 215.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5288
Linear Formula (C5H4N)CN
Quantity 5 g
Beilstein 22, 36
Solubility Information Solubility in water: immiscible.
Formula Weight 104.11
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material 2-Cyanopyridine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-880-3
TSCA TSCA

RUO – Research Use Only