missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyridine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036402
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54689MD 250 mg
AAH5468903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54689MD Supplier Thermo Scientific Chemicals Supplier No. H54689MD

CAS: 741709-63-7 | C12H15BN2O2 | 230.07 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is used in suzuki reactions.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).

Chemical Identifiers

CAS 741709-63-7
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.07
MDL Number MFCD06657825
InChI Key IXTBQKLZPOYJFJ-UHFFFAOYSA-N
Synonym 2-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyano pyridine-5-boronic acid pinacol ester, 2-cyanopyridin-5-yl boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyanopyridine-5-boronic acid pinacole ester, 6-cyanopyridine-3-boronic acid pinacol ester, 2-pyridinecarbonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarbonitrile
PubChem CID 16414188
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C#N

Specifications

Melting Point 105°C to 106°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 230.07
Percent Purity 96%
Chemical Name or Material 2-Cyanopyridine-5-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only