missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoisonicotinic Acid 98.0+%, TCI America™

Catalog No. C3396200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3396200MG 200 mg
C33961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3396200MG Supplier TCI America Supplier No. C3396200MG
Only null left

Chemical Identifiers

CAS 161233-97-2
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD03411729
InChI Key MPSVJNPESHZCIB-UHFFFAOYSA-N
Synonym 2-Cyano-4-pyridinecarboxylic Acid
PubChem CID 2762513
IUPAC Name 2-cyanopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)C#N

Specifications

Melting Point 335°C
Color White-Yellow
Quantity 200 mg
Formula Weight 148.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanoisonicotinic Acid