missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyltriethoxysilane 98.0+%, TCI America™

Catalog No. C125925ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C125925ML 25 mL
C1259250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C125925ML Supplier TCI America Supplier No. C125925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 919-31-3
Molecular Formula C9H19NO3Si
Molecular Weight (g/mol) 217.34
MDL Number MFCD00019851
InChI Key GBQYMXVQHATSCC-UHFFFAOYSA-N
Synonym 3-triethoxysilyl propionitrile, 2-cyanoethyl triethoxysilane, 2-cyanoethyltriethoxysilane, propanenitrile, 3-triethoxysilyl, beta-cyanoethyltriethoxysilane, 3-triethoxysilyl propanenitrile, propionitrile, 3-triethoxysilyl, silane, 2-cyanoethyl triethoxy, triethoxy-2-kyanethylsilan, unii-0i8r6d195f
PubChem CID 13522
IUPAC Name 3-triethoxysilylpropanenitrile
SMILES CCO[Si](CCC#N)(OCC)OCC

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 mL
Formula Weight 217.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyanoethyltriethoxysilane
RTECSNumber VV2750000
TSCA Yes