missing translation for 'onlineSavingsMsg'
Learn More

(2-Cyanoethyl)triethoxysilane, 97%

Catalog No. AAL0354314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0354314 25 g
AAL0354322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0354314 Supplier Thermo Scientific Chemicals Supplier No. L0354314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 919-31-3 | C9H19NO3Si | 217.34 g/mol

(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Cyanoethyl)triethoxysilane is an ethoxylated silicate that has been used as a coating for aluminum and quartz.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 919-31-3
Molecular Formula C9H19NO3Si
Molecular Weight (g/mol) 217.34
MDL Number MFCD00019851
InChI Key GBQYMXVQHATSCC-UHFFFAOYSA-N
Synonym 3-triethoxysilyl propionitrile, 2-cyanoethyl triethoxysilane, 2-cyanoethyltriethoxysilane, propanenitrile, 3-triethoxysilyl, beta-cyanoethyltriethoxysilane, 3-triethoxysilyl propanenitrile, propionitrile, 3-triethoxysilyl, silane, 2-cyanoethyl triethoxy, triethoxy-2-kyanethylsilan, unii-0i8r6d195f
PubChem CID 13522
IUPAC Name 3-triethoxysilylpropanenitrile
SMILES CCO[Si](CCC#N)(OCC)OCC

Specifications

Boiling Point 224°C to 225°C
Quantity 25 g
Beilstein 1776392
Flash Point 100°C (212°F)
Solubility Information Hydrolyzes in water.
Formula Weight 217.34
Percent Purity 97%
Refractive Index 1.413
Sensitivity Moisture sensitive
Density 0.98
Chemical Name or Material (2-Cyanoethyl)triethoxysilane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-050-5
RTECSNumber VV2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only