missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. C18115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18115G 5 g
C18111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18115G Supplier TCI America Supplier No. C18115G

Chemical Identifiers

CAS 69360-26-5
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
InChI Key NQAYCMBZPAARNO-UHFFFAOYSA-N
Synonym 2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7
PubChem CID 522347
IUPAC Name 2-cyanobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl

Specifications

Melting Point 68°C
Quantity 5 g
UN Number 2928
Formula Weight 201.62
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanobenzenesulfonyl Chloride