missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzenesulfonyl chloride, 97%

Catalog No. p-7023021
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1604206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1604206 Supplier Thermo Scientific Chemicals Supplier No. A1604206

CAS: 69360-26-5 | C7H4ClNO2S | 201.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69360-26-5
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
MDL Number MFCD00052341
InChI Key NQAYCMBZPAARNO-UHFFFAOYSA-N
Synonym 2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7
PubChem CID 522347
IUPAC Name 2-cyanobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl

Specifications

Melting Point 65°C to 69°C
Quantity 5 g
UN Number UN3261
Beilstein 2721588
Sensitivity Moisture sensitive
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 2-Cyanobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only