Learn More
2-Cyanobenzeneboronic acid pinacol ester, 98%
CAS: 214360-48-2 | C13H16BNO2 | 229.086 g/mol
$220.66 - $734.37
Chemical Identifiers
| CAS | 214360-48-2 |
|---|---|
| Molecular Formula | C13H16BNO2 |
| Molecular Weight (g/mol) | 229.086 |
| MDL Number | MFCD04038747 |
| InChI Key | SKQNWSBNAIOCOC-UHFFFAOYSA-N |
| Synonym | 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p |
| PubChem CID | 3570229 |
| IUPAC Name | 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2746906
|
Thermo Scientific Chemicals
H2746906 |
5 g |
Each for $220.66
|
|
|||||
|
AAH2746914
|
Thermo Scientific Chemicals
H2746914 |
25 g |
Each for $734.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 214360-48-2 | |
| 229.086 | |
| SKQNWSBNAIOCOC-UHFFFAOYSA-N | |
| 3570229 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N |
| C13H16BNO2 | |
| MFCD04038747 | |
| 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p | |
| 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
Specifications
| 214360-48-2 | |
| C13H16BNO2 | |
| 5 g | |
| SKQNWSBNAIOCOC-UHFFFAOYSA-N | |
| 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile | |
| 3570229 | |
| 98% |
| 88°C to 90°C | |
| MFCD04038747 | |
| 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N | |
| 229.086 | |
| 229.08 | |
| 2-Cyanobenzeneboronic acid pinacol ester |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only