missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzeneboronic acid pinacol ester, 98%

Catalog No. AAH2746914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2746914 25 g
AAH2746906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2746914 Supplier Thermo Scientific Chemicals Supplier No. H2746914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 214360-48-2 | C13H16BNO2 | 229.086 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214360-48-2
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.086
MDL Number MFCD04038747
InChI Key SKQNWSBNAIOCOC-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p
PubChem CID 3570229
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N

Specifications

Melting Point 88°C to 90°C
Quantity 25 g
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Cyanobenzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only