missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoacetanilide 98.0+%, TCI America™

Catalog No. p-7125707
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28941G 1 g
C28945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28941G Supplier TCI America Supplier No. C28941G
Only null left

Chemical Identifiers

CAS 621-03-4
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00019840
InChI Key XCTQPMCULSZKLT-UHFFFAOYSA-N
Synonym 2-cyanoacetanilide, acetamide, 2-cyano-n-phenyl, n-phenylcyanoacetamide, 2-cyano-n-phenyl-acetamide, alpha-cyanoacetanilide, acetanilide, 2-cyano, cyanoacetanilide, cyanoacetic acid anilide, 2-cyano-acetanilide, n-phenyl cyanoacetamide
PubChem CID 69296
IUPAC Name 2-cyano-N-phenylacetamide
SMILES O=C(CC#N)NC1=CC=CC=C1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 1 g
Formula Weight 160.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanoacetanilide