missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-N,N-dimethylacetamide 98.0+%, TCI America™

Catalog No. C17935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17935G 5 g
C179325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17935G Supplier TCI America Supplier No. C17935G

Chemical Identifiers

CAS 7391-40-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.13
MDL Number MFCD00051623
InChI Key MATJPVGBSAQWAC-UHFFFAOYSA-N
Synonym n,n-dimethylcyanoacetamide, acetamide, 2-cyano-n,n-dimethyl, n,n-dimethyl cyanoacetamide, n,n-dimethyl-2-cyanoacetamide, acmc-209osw, n,n-dimethylcyanoacetate, 2-cyano-n,n-dimethyl-acetamide, 2-cyano-n,n-dimethyl-ethanamide, acetamide,2-cyano-n,n-dimethyl, cyanoacetic acid n,n-dimethylamide
PubChem CID 251143
IUPAC Name 2-cyano-N,N-dimethylacetamide
SMILES CN(C)C(=O)CC#N

Specifications

Melting Point 66°C
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-N,N-dimethylacetamide