missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-N,N-diethylacetamide 98.0+%, TCI America™

Catalog No. c28225g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28225G Supplier TCI America Supplier No. C28225G

Specifications

CAS 26391-06-0
Color Yellow
Boiling Point 114°C
Molecular Formula C7H12N2O
Quantity 5 g
MDL Number MFCD01332383
InChI Key RYSHIRFTLKZVIH-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)CC#N
IUPAC Name 2-cyano-N,N-diethylacetamide
Molecular Weight (g/mol) 140.19
PubChem CID 3854515
ChEBI CHEBI:48386
Formula Weight 140.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyano-N,N-diethylacetamide
Show More Show Less
TSCA No
Recommended Storage Refrigerator