missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methylpyridine, 98%

Catalog No. p-7055541
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6414806 5 g
AAH6414814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6414806 Supplier Thermo Scientific Chemicals Supplier No. H6414806
Only null left

CAS: 1620-75-3 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1620-75-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00190586
InChI Key CMADFEQMYFNYCF-UHFFFAOYSA-N
Synonym 6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril
PubChem CID 74182
IUPAC Name 6-methylpyridine-2-carbonitrile
SMILES CC1=CC=CC(=N1)C#N

Specifications

Melting Point 70°C to 74°C
Color White
Quantity 5 g
UN Number UN3439
Beilstein 111107
Formula Weight 118.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Cyano-6-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 216-588-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only