missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methylpyridine, 97%

Catalog No. AC425920050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC425920050 5 g
AC425920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425920050 Supplier Thermo Scientific Chemicals Supplier No. 425920050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1620-75-3 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1620-75-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00190586
InChI Key CMADFEQMYFNYCF-UHFFFAOYSA-N
Synonym 6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril
PubChem CID 74182
IUPAC Name 6-methylpyridine-2-carbonitrile
SMILES CC1=CC=CC(=N1)C#N

Specifications

Melting Point 70.0°C to 74.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 2-Cyano-6-methylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 216-588-9

RUO – Research Use Only