missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-methoxybenzothiazole 97.0+%, TCI America™

Catalog No. C11761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11761G Supplier TCI America Supplier No. C11761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 943-03-3
Molecular Formula C9H6N2OS
Molecular Weight (g/mol) 190.22
MDL Number MFCD00010537
InChI Key DEWDWBYQOFXKIH-UHFFFAOYSA-N
Synonym 2-cyano-6-methoxybenzothiazole, 6-methoxybenzo d thiazole-2-carbonitrile, 6-methoxy-2-benzothiazolecarbonitrile, 6-methoxybenzothiazole-2-carbonitrile, 6-methoxy-benzothiazole-2-carbonitrile, 6-methoxy-2-cyano-benzothiazole, 2-benzothiazolecarbonitrile, 6-methoxy, pubchem9766, acmc-209rr8, ksc486m5p
PubChem CID 342109
IUPAC Name 6-methoxy-1,3-benzothiazole-2-carbonitrile
SMILES COC1=CC2=C(C=C1)N=C(S2)C#N

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 190.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-6-methoxybenzothiazole