missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-(trifluoromethyl)pyridine, 98%

Catalog No. p-7055812
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6448603 1 g
AAH6448606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6448603 Supplier Thermo Scientific Chemicals Supplier No. H6448603
Only null left

CAS: 95727-86-9 | C7H3F3N2 | 172.11 g/mol

It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 95727-86-9
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD01001119
InChI Key WDSCJULUXJSJOX-UHFFFAOYSA-N
Synonym 5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
PubChem CID 1535372
IUPAC Name 5-(trifluoromethyl)pyridine-2-carbonitrile
SMILES FC(F)(F)C1=CN=C(C=C1)C#N

Specifications

Quantity 1 g
UN Number UN3439
Beilstein 13339826
Solubility Information Slightly soluble in water.
Formula Weight 172.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Cyano-5-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only