missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. C3126200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3126200MG 200 mg
C31261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3126200MG Supplier TCI America Supplier No. C3126200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 741709-63-7
Molecular Formula C12H15BN2O2
Molecular Weight (g/mol) 230.07
MDL Number MFCD06657825
InChI Key IXTBQKLZPOYJFJ-UHFFFAOYSA-N
Synonym 2-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyano pyridine-5-boronic acid pinacol ester, 2-cyanopyridin-5-yl boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carbonitrile, 2-cyanopyridine-5-boronic acid pinacole ester, 6-cyanopyridine-3-boronic acid pinacol ester, 2-pyridinecarbonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarbonitrile
PubChem CID 16414188
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C#N

Specifications

Melting Point 112°C
Quantity 200 mg
Formula Weight 230.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine