missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4-methylpyridine 98.0+%, TCI America™

Catalog No. p-7125551
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C26035G 5 g
C260325G 25 g
2 options

Catalog No. C26035G

Supplier: TCI America C26035G

Only null left

Chemical Identifiers

CAS 1620-76-4
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00128868
InChI Key LQAWSWUFSHYCHP-UHFFFAOYSA-N
Synonym 2-cyano-4-methylpyridine, 4-methylpicolinonitrile, 4-methyl-2-pyridinecarbonitrile, 2-cyano-4-picoline, 2-pyridinecarbonitrile, 4-methyl, pubchem21387, acmc-209dnq, 2-cyano-4methylpyridine, 2-cyano 4-methylpyridine, 4-methyl-2-cyanopyridine
PubChem CID 88739
IUPAC Name 4-methylpyridine-2-carbonitrile
SMILES CC1=CC(=NC=C1)C#N

Specifications

Melting Point 88°C
Boiling Point 148°C
Quantity 5 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-4-methylpyridine