missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4-methylpyridine, 98%

Catalog No. AAL1714806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1714806 5 g
AAL1714803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1714806 Supplier Thermo Scientific Chemicals Supplier No. L1714806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1620-76-4 | C7H6N2 | 118.139 g/mol

2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1620-76-4
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00128868
InChI Key LQAWSWUFSHYCHP-UHFFFAOYSA-N
Synonym 2-cyano-4-methylpyridine, 4-methylpicolinonitrile, 4-methyl-2-pyridinecarbonitrile, 2-cyano-4-picoline, 2-pyridinecarbonitrile, 4-methyl, pubchem21387, acmc-209dnq, 2-cyano-4methylpyridine, 2-cyano 4-methylpyridine, 4-methyl-2-cyanopyridine
PubChem CID 88739
IUPAC Name 4-methylpyridine-2-carbonitrile
SMILES CC1=CC(=NC=C1)C#N

Specifications

Melting Point 85°C to 88°C
Boiling Point 145°C to 148°C (38 mmHg)
Odor Characteristic
Quantity 5 g
Beilstein 110753
Solubility Information Slightly soluble in water.
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 2-Cyano-4-methylpyridine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only