missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
$127.10 - $127.10
Chemical Identifiers
| CAS | 461451-63-8 |
|---|---|
| Molecular Formula | C13H15BFNO2 |
| Molecular Weight (g/mol) | 247.076 |
| MDL Number | MFCD09260444 |
| InChI Key | IMEIHRCZDURBEC-UHFFFAOYSA-N |
| Synonym | 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronic acid pinacol ester, 2-cyano-4-fluorobenzeneboronic acid pinacol ester, 5-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronicacidpinacolester, 2-cyano-4-fluorophenyl boronic acid pinacol ester, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 5-fluoranyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, 5-fluoro-2-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile |
| PubChem CID | 22180790 |
| IUPAC Name | 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH62729MD
|
Thermo Scientific Chemicals
H62729MD |
250 mg |
Each for $127.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 461451-63-8 | |
| 247.076 | |
| IMEIHRCZDURBEC-UHFFFAOYSA-N | |
| 22180790 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C#N |
| C13H15BFNO2 | |
| MFCD09260444 | |
| 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronic acid pinacol ester, 2-cyano-4-fluorobenzeneboronic acid pinacol ester, 5-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronicacidpinacolester, 2-cyano-4-fluorophenyl boronic acid pinacol ester, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 5-fluoranyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, 5-fluoro-2-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile | |
| 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
Specifications
| 461451-63-8 | |
| MFCD09260444 | |
| 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronic acid pinacol ester, 2-cyano-4-fluorobenzeneboronic acid pinacol ester, 5-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyano-4-fluorophenylboronicacidpinacolester, 2-cyano-4-fluorophenyl boronic acid pinacol ester, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 5-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 5-fluoranyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, 5-fluoro-2-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)F)C#N | |
| 247.076 | |
| 247.07 | |
| Solid |
| C13H15BFNO2 | |
| 250 mg | |
| IMEIHRCZDURBEC-UHFFFAOYSA-N | |
| 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile | |
| 22180790 | |
| 96% | |
| 2-Cyano-4-fluorobenzeneboronic acid pinacol ester |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only