missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-3-methylpyridine 98.0+%, TCI America™
$29.83 - $68.97
Chemical Identifiers
| CAS | 20970-75-6 |
|---|---|
| Molecular Formula | C7H6N2 |
| Molecular Weight (g/mol) | 118.139 |
| MDL Number | MFCD00190585 |
| InChI Key | WBXZCDIZXWDPBL-UHFFFAOYSA-N |
| Synonym | 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline |
| PubChem CID | 819928 |
| IUPAC Name | 3-methylpyridine-2-carbonitrile |
| SMILES | CC1=C(N=CC=C1)C#N |
Chemical Identifiers
| 20970-75-6 | |
| 118.139 | |
| WBXZCDIZXWDPBL-UHFFFAOYSA-N | |
| 819928 | |
| CC1=C(N=CC=C1)C#N |
| C7H6N2 | |
| MFCD00190585 | |
| 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline | |
| 3-methylpyridine-2-carbonitrile |
Specifications
| 20970-75-6 | |
| White-Yellow | |
| C7H6N2 | |
| 5 g | |
| 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline | |
| CC1=C(N=CC=C1)C#N | |
| 118.139 | |
| 118.14 | |
| Crystalline Powder |
| 86°C | |
| 142°C | |
| MFCD00190585 | |
| 3439 | |
| WBXZCDIZXWDPBL-UHFFFAOYSA-N | |
| 3-methylpyridine-2-carbonitrile | |
| 819928 | |
| ≥98.0% (GC,T) | |
| 2-Cyano-3-methylpyridine |
Safety and Handling
TSCA : No