missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-fluoropyridine 98.0+%, TCI America™

Catalog No. C26551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26551G 1 g
C26555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26551G Supplier TCI America Supplier No. C26551G

Chemical Identifiers

CAS 97509-75-6
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD06797501
InChI Key VZFPSCNTFBJZHB-UHFFFAOYSA-N
Synonym 2-cyano-3-fluoropyridine, 3-fluoropicolinonitrile, 3-fluoro-2-pyridinecarbonitrile, 3-fluoro-2-cyanopyridine, 3-fluoro-2-pyridinenitrile, 3-fluoro-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-fluoro, fluorocyanopyridine, pubchem16434, acmc-209s8i
PubChem CID 7060408
IUPAC Name 3-fluoropyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)F

Specifications

Melting Point 32°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 122.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Cyano-3-fluoropyridine