Learn More
Cytiva 2 CHR Cellulose Blotting Paper
Catalog No. 09927699
Change view
Click to view available options
Dimensions (L x W):
58 x 60 cm (22.8 x 23.6 in.)
Catalog No. Dimensions (L x W)
09927699 58 x 60 cm (22.8 x 23.6 in.)
1 options
Catalog No. 09927699 Supplier Cytiva Supplier No. 3002911
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Smooth surface. Particularly recommended for optical or radiometric scanning
Thickness 0.18mm. Flow rate 115mm/30 min. Slower than 1 Chr for higher resolution applications. Smooth surface. Particularly recommended for optical or radiometric scanning.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
Specifications
Grade Grade 2 Chr
Description Grade 2 Chr cellulose chromatography sheets is a 0.18 mm paper for higher resolution applications.
Dimensions (L x W) 58 x 60 cm (22.8 x 23.6 in.)
Material Cellulose