missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorotritanol, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 66774-02-5 |
| Flash Point | Not Applicable |
| Molecular Formula | C19H15ClO |
| Linear Formula | C19H15ClO |
| Quantity | 25 mg |
| Synonym | (o-Chlorophenyl)diphenylmethanol |
| Formula Weight | 294.77 |
| Grade | British Pharmacopoeia (BP) Reference Standard |