missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorotoluene 98.0+%, TCI America™
$76.43 - $119.30
Chemical Identifiers
| CAS | 95-49-8 |
|---|---|
| Molecular Formula | C7H7Cl |
| Molecular Weight (g/mol) | 126.58 |
| MDL Number | MFCD00000562 |
| InChI Key | IBSQPLPBRSHTTG-UHFFFAOYSA-N |
| Synonym | 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene |
| PubChem CID | 7238 |
| IUPAC Name | 1-chloro-2-methylbenzene |
| SMILES | CC1=CC=CC=C1Cl |
Chemical Identifiers
| 95-49-8 | |
| 126.58 | |
| IBSQPLPBRSHTTG-UHFFFAOYSA-N | |
| 7238 | |
| CC1=CC=CC=C1Cl |
| C7H7Cl | |
| MFCD00000562 | |
| 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene | |
| 1-chloro-2-methylbenzene |
Specifications
| -35°C | |
| Colorless | |
| 25 g | |
| C7H7Cl | |
| 2238 | |
| IBSQPLPBRSHTTG-UHFFFAOYSA-N | |
| 1-chloro-2-methylbenzene | |
| 7238 | |
| ≥98.0% (GC) |
| 95-49-8 | |
| 159°C | |
| Clear Liquid at 20°C | |
| MFCD00000562 | |
| 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene | |
| CC1=CC=CC=C1Cl | |
| 126.58 | |
| 126.58 | |
| 2-Chlorotoluene |
Safety and Handling
EINECSNumber : (3)-0039
RTECSNumber : XS9000000
TSCA : Yes