missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene 98.0+%, TCI America™

Catalog No. C029525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C029525G 25 g
C0295500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C029525G Supplier TCI America Supplier No. C029525G

Chemical Identifiers

CAS 95-49-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
Synonym 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Melting Point -35°C
Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 2238
Boiling Point 159°C
Formula Weight 126.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chlorotoluene
EINECSNumber (3)-0039
RTECSNumber XS9000000
TSCA Yes