missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%

CAS: 95-49-8 | C7H7Cl | 126.58 g/mol

Supplier:  Thermo Scientific Chemicals B235960B

 View more versions of this product

Catalog No. AAB235960B


Only null left
Explore available promotions

Description

Description

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Specifications

2-Chlorotoluene
95-49-8
157°C to 159°C
1.525
MFCD00000562
1904175
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
IBSQPLPBRSHTTG-UHFFFAOYSA-N
1-chloro-2-methylbenzene
7238
99%
-36°C
1.082
1000 g
C7H7Cl
UN2238
14,2171
Slightly soluble in water.
CC1=CC=CC=C1Cl
126.58
126.59

Chemical Identifiers

95-49-8
126.58
IBSQPLPBRSHTTG-UHFFFAOYSA-N
7238
CC1=CC=CC=C1Cl
C7H7Cl
MFCD00000562
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
1-chloro-2-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.

P201-P202-P210-P233-P240-P241-P242-P243-P261-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c

H226-H332-H361d

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROTOLUENES

EINECSNumber : 202-424-3

RTECSNumber : XS9000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only