missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%

Catalog No. AAB2359622
Change view
Click to view available options
Quantity:
100 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2359622 100 g
AAB235960B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2359622 Supplier Thermo Scientific Chemicals Supplier No. B2359622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-49-8 | C7H7Cl | 126.58 g/mol

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 95-49-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
Synonym 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Melting Point -36°C
Density 1.082
Quantity 100 g
Refractive Index 1.525
UN Number UN2238
Beilstein 1904175
Merck Index 14,2171
Boiling Point 157°C to 159°C
Solubility Information Slightly soluble in water.
Formula Weight 126.59
Percent Purity 99%
Chemical Name or Material 2-Chlorotoluene
Show More Show Less
Hazard Category H226-H332-H361d
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROTOLUENES
EINECSNumber 202-424-3
RTECSNumber XS9000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only