missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophenol, 98%, Thermo Scientific™

Catalog No. AAA1201406
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1201406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1201406 Supplier Thermo Scientific Chemicals Supplier No. A1201406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 6320-03-2
Density 1.277
Boiling Point 108°C to 110°C (30mmHg)
Flash Point 88°C (190°F)
Molecular Formula C6H5ClS
Refractive Index 1.604
Quantity 5g
MDL Number MFCD00004830
UN Number UN1760
Beilstein 1209665
Sensitivity Air Sensitive
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)S)Cl
IUPAC Name 2-chlorobenzenethiol
Molecular Weight (g/mol) 144.62
PubChem CID 80599
Formula Weight 144.62
Percent Purity 98%
Chemical Name or Material 2-Chlorothiophenol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 228-667-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only