missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorothiazole 98.0+%, TCI America™
$78.93 - $234.87
Chemical Identifiers
| CAS | 3034-52-4 |
|---|---|
| Molecular Formula | C3H2ClNS |
| Molecular Weight (g/mol) | 119.566 |
| MDL Number | MFCD00210701 |
| InChI Key | KLEYVGWAORGTIT-UHFFFAOYSA-N |
| Synonym | 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 |
| PubChem CID | 76429 |
| ChEBI | CHEBI:39187 |
| IUPAC Name | 2-chloro-1,3-thiazole |
| SMILES | C1=CSC(=N1)Cl |
Chemical Identifiers
| 3034-52-4 | |
| 119.566 | |
| KLEYVGWAORGTIT-UHFFFAOYSA-N | |
| 76429 | |
| 2-chloro-1,3-thiazole |
| C3H2ClNS | |
| MFCD00210701 | |
| 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
| CHEBI:39187 | |
| C1=CSC(=N1)Cl |
Specifications
| 3034-52-4 | |
| 145°C | |
| MFCD00210701 | |
| 1993 | |
| KLEYVGWAORGTIT-UHFFFAOYSA-N | |
| 2-chloro-1,3-thiazole | |
| 76429 | |
| 119.57 | |
| Liquid |
| Yellow | |
| C3H2ClNS | |
| 5 g | |
| 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
| C1=CSC(=N1)Cl | |
| 119.566 | |
| CHEBI:39187 | |
| ≥98.0% (GC) | |
| 2-Chlorothiazole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator