missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiazole, 97%

Catalog No. AAH5015506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5015506 5 g
AAH5015503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5015506 Supplier Thermo Scientific Chemicals Supplier No. H5015506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3034-52-4 | C3H2ClNS | 119.566 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-52-4
Molecular Formula C3H2ClNS
Molecular Weight (g/mol) 119.566
MDL Number MFCD00210701
InChI Key KLEYVGWAORGTIT-UHFFFAOYSA-N
Synonym 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569
PubChem CID 76429
ChEBI CHEBI:39187
IUPAC Name 2-chloro-1,3-thiazole
SMILES C1=CSC(=N1)Cl

Specifications

Quantity 5 g
Formula Weight 119.57
Percent Purity 97%
Chemical Name or Material 2-Chlorothiazole
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-228-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only