Learn More
2-Chlorothiazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 457260050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-Chlorothiazole | |
| 1.3780g/mL | |
| 54°C | |
| 96% min. (GC) | |
| 5g | |
| 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
| C1=CSC(=N1)Cl | |
| 119.57 | |
| CHEBI:39187 | |
| 97% |
| 3034-52-4 | |
| 145.0°C to 146.0°C | |
| Authentic | |
| C3H2ClNS | |
| 1.378 | |
| KLEYVGWAORGTIT-UHFFFAOYSA-N | |
| 2-chloro-1,3-thiazole | |
| 76429 | |
| 119.57 |
Chemical Identifiers
| 3034-52-4 | |
| 119.57 | |
| 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
| CHEBI:39187 | |
| C1=CSC(=N1)Cl |
| C3H2ClNS | |
| KLEYVGWAORGTIT-UHFFFAOYSA-N | |
| 76429 | |
| 2-chloro-1,3-thiazole |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Warning
EINECSNumber : 221-228-9
RUO â Research Use Only