missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroquinoline 98.0+%, TCI America™

Catalog No. p-7124332
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C02825G 5 g
C028225G 25 g
2 options

Catalog No. C02825G

Supplier: TCI America C02825G

Only null left

Chemical Identifiers

CAS 612-62-4
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00006741
InChI Key OFUFXTHGZWIDDB-UHFFFAOYSA-N
Synonym 2-chloro-quinoline, quinoline, 2-chloro, chloroquinoline, unii-ux7rin8gew, quinoline, chloro, ccris 3977, ux7rin8gew, chioroquinoline, 2-chloro quinoline, pubchem7580
PubChem CID 11928
IUPAC Name 2-chloroquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)Cl

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 267°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloroquinoline
RTECSNumber VB2320000
TSCA Yes
Recommended Storage Refrigerator