missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroquinoline-4-carboxylic Acid 97.0+%, TCI America™
$288.94 - $1060.65
Chemical Identifiers
| CAS | 5467-57-2 |
|---|---|
| Molecular Formula | C10H5ClNO2 |
| Molecular Weight (g/mol) | 206.61 |
| MDL Number | MFCD00023940 |
| InChI Key | ICNCOMYUODLTAI-UHFFFAOYSA-M |
| PubChem CID | 230582 |
| IUPAC Name | 2-chloroquinoline-4-carboxylate |
| SMILES | [O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1 |
Chemical Identifiers
| 5467-57-2 | |
| 206.61 | |
| ICNCOMYUODLTAI-UHFFFAOYSA-M | |
| 2-chloroquinoline-4-carboxylate |
| C10H5ClNO2 | |
| MFCD00023940 | |
| 230582 | |
| [O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1 |
Specifications
| 5467-57-2 | |
| C10H5ClNO2 | |
| 5 g | |
| [O-]C(=O)C1=C2C=CC=CC2=NC(Cl)=C1 | |
| 206.61 | |
| 207.61 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00023940 | |
| ICNCOMYUODLTAI-UHFFFAOYSA-M | |
| 2-chloroquinoline-4-carboxylate | |
| 230582 | |
| ≥97.0% (HPLC,T) | |
| 2-Chloroquinoline-4-carboxylic Acid |
Safety and Handling
TSCA : No