missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-5-carboxylic acid, 97%

Catalog No. AC429470010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429470010 1 g Glass bottle
AC429470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429470010 Supplier Thermo Scientific Chemicals Supplier No. 429470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 374068-01-6 | C5H3ClN2O2 | 158.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374068-01-6
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD06739036
InChI Key DUCXUPKLVVSJKA-UHFFFAOYSA-N
Synonym 2-chloro-pyrimidine-5-carboxylic acid, 2-chloro-5-carboxypyrimidine, 5-carboxy-2-chloropyrimidine, 5-pyrimidinecarboxylicacid, 2-chloro, 2-chloro-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-chloro, 2-chloropyrimidine-5-carboxylicacid, acmc-1aig3, 2-chloropyrimidine 5-carboxylic acid, 2-chloro-5-pyrimidine carboxylic acid
PubChem CID 11094906
IUPAC Name 2-chloropyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=C(Cl)N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 125.0°C to 127.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 158.54
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Chloropyrimidine-5-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only