missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

$1004.45 - $1004.45

Chemical Identifiers

CAS 1003845-08-6
Molecular Formula C10H14BClN2O2
Molecular Weight (g/mol) 240.49
MDL Number MFCD11856048
InChI Key VLAPDEKXZLRRKV-UHFFFAOYSA-N
Synonym 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 51341943
IUPAC Name 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6405206
View Documents
Thermo Scientific Chemicals
H6405206
5 g
Each for $1,004.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1003845-08-6
240.49
VLAPDEKXZLRRKV-UHFFFAOYSA-N
51341943
CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
C10H14BClN2O2
MFCD11856048
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Specifications

1003845-08-6
MFCD11856048
15622160
VLAPDEKXZLRRKV-UHFFFAOYSA-N
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
51341943
98%
2-Chloropyrimidine-5-boronic acid pinacol ester
C10H14BClN2O2
5 g
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
240.49
240.49
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only