missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine 98.0+%, TCI America™

Catalog No. p-7124329
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C027925G 25 g
C0279100G 100 g
C0279500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C027925G Supplier TCI America Supplier No. C027925G
Only null left

Chemical Identifiers

CAS 109-09-1
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.544
MDL Number MFCD00006228
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

Color Yellow
pH 6 to 7 (27g/L H2O 20°C)
Boiling Point 170°C
Quantity 25 g
UN Number 2822
Formula Weight 113.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloropyridine
EINECSNumber (5)-0720
RTECSNumber US5950000
TSCA Yes