missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine N-oxide hydrochloride, 97%

Catalog No. p-7029778
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2728006 5 g
AAH2728014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2728006 Supplier Thermo Scientific Chemicals Supplier No. H2728006

CAS: 20295-64-1 | C5H5Cl2NO | 166.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20295-64-1
Molecular Formula C5H5Cl2NO
Molecular Weight (g/mol) 166.001
MDL Number MFCD00007913
InChI Key GRZNODNSNCXOHE-UHFFFAOYSA-N
Synonym 2-chloropyridine n-oxide hydrochloride, 2-chloropyridine-n-oxide hydrochloride, 2-chloropyridine 1-oxide hydrochloride, chloride 2-chloropyridinium 1-oxide, 2-chloropyridine n-oxide hcl, pyridine, 2-chloro-, 1-oxide, hydrochloride, 2-chloropyridin-1-ium-1-olate hydrochloride, 2-chlorpyridin-1-oxidhydrochlorid, acmc-1ccui, 2-chloropyridine n-oxide.hcl
PubChem CID 88478
IUPAC Name 2-chloro-1-oxidopyridin-1-ium;hydrochloride
SMILES C1=CC=[N+](C(=C1)Cl)[O-].Cl

Specifications

Melting Point 138°C to 142°C
Quantity 5 g
Formula Weight 166.01
Percent Purity 97%
Chemical Name or Material 2-Chloropyridine N-oxide hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-712-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only