missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine, 99%

Catalog No. AAA1486622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1486622 100 g
AAA1486636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1486622 Supplier Thermo Scientific Chemicals Supplier No. A1486622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-09-1 | C5H4ClN | 113.544 g/mol

2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

Solubility
Soluble in water (27g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 109-09-1
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.544
MDL Number MFCD00006228
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

Melting Point -46°C
Density 1.208
pH 6 to 7
Boiling Point 167°C to 170°C
Flash Point 65°C (149°F)
Refractive Index 1.532
Quantity 100 g
UN Number UN2822
Beilstein 105788
Solubility Information Soluble in water (27g/L).
Formula Weight 113.55
Percent Purity 99%
Chemical Name or Material 2-Chloropyridine
Show More Show Less
Hazard Category H227-H302-H310+H330-H315-H318-H373
Hazard Statement GHS H Statement
H301-H310-H330-H318-H227-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P280-P284-P301+P312-P302+P350-P304+P340-P305+P351+P338-P310-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-CHLOROPYRIDINE
EINECSNumber 203-646-3
RTECSNumber US5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only