missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
$80.16 - $80.16
Specifications
| CAS | 444120-91-6 |
|---|---|
| Melting Point | 167°C |
| Color | White-Yellow |
| Molecular Formula | C5H5BClNO2 |
| MDL Number | MFCD03094998 |
Specifications
| 444120-91-6 | |
| White-Yellow | |
| MFCD03094998 | |
| (6-chloropyridin-3-yl)boronic acid | |
| 3845034 | |
| Crystalline Powder |
| 167°C | |
| C5H5BClNO2 | |
| WPAPNCXMYWRTTL-UHFFFAOYSA-N | |
| 157.36 | |
| 157.36 |