missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C22881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22881G Supplier TCI America Supplier No. C22881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Chloropyridine-5-boronic Acid (contains varying amounts of Anhydride)
CAS 444120-91-6
Melting Point 167°C
Color White-Yellow
Molecular Formula C5H5BClNO2
Quantity 1 g
MDL Number MFCD03094998
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(Cl)N=C1
IUPAC Name (6-chloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 157.36
PubChem CID 3845034
Formula Weight 157.36
Percent Purity 96.0 to 113.0%
Physical Form Crystalline Powder
Show More Show Less