missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine-3-carboxaldehyde, 98%

Catalog No. p-7035676
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5339903 1 g
AAH5339906 5 g
AAH5339914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5339903 Supplier Thermo Scientific Chemicals Supplier No. H5339903

CAS: 36404-88-3 | C6H4ClNO | 141.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36404-88-3
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD01315308
InChI Key KHPAGGHFIDLUMB-UHFFFAOYSA-N
Synonym 2-chloro-3-formylpyridine, 2-chloronicotinaldehyde, 2-chloro-3-pyridinecarboxaldehyde, 2-chloro-pyridine-3-carbaldehyde, chloro-2-formyl-3-pyridine, 2-chloropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-chloro, 2-chloro-3-pyridinecarbaldehyde, 2-chloro-3-pyridinecarboxyaldehyde, 2-chloro-pyridin-3-carbaldehyde
PubChem CID 737607
IUPAC Name 2-chloropyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Cl)C=O

Specifications

Melting Point 50°C to 54°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 141.56
Percent Purity 98%
Chemical Name or Material 2-Chloropyridine-3-carboxaldehyde
Hazard Category H302+H312+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only