missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C25601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25601G Supplier TCI America Supplier No. C25601G

Specifications

Chemical Name or Material 2-Chloropyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 381248-04-0
Melting Point 160°C
Color White-Yellow
Molecular Formula C5H5BClNO2
Quantity 1 g
MDL Number MFCD03094997
Synonym 2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
InChI Key VRDAOVQZVXYRNH-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CN=C1Cl
IUPAC Name (2-chloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 157.36
PubChem CID 2762704
Formula Weight 157.36
Physical Form Crystalline Powder
Show More Show Less