missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropyrazine, 98%
Description
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.
Solubility
Soluble in Methanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.
Solubility
Soluble in Methanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Density | 1.284 |
| Boiling Point | 153°C to 154°C |
| Flash Point | 57°C (134°F) |
| Refractive Index | 1.537 |
| Quantity | 5 g |
| UN Number | UN1993 |
| Beilstein | 107019 |
| Solubility Information | Soluble in Methanol |
| Formula Weight | 114.54 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 238-517-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only