missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropropiophenone 95.0+%, TCI America™
$257.24 - $902.21
Chemical Identifiers
| CAS | 6084-17-9 |
|---|---|
| Molecular Formula | C9H9ClO |
| Molecular Weight (g/mol) | 168.62 |
| MDL Number | MFCD03844782 |
| InChI Key | AXCPQHPNAZONTH-UHFFFAOYNA-N |
| Synonym | 2-Chloro-1-phenyl-1-propanone |
| PubChem CID | 221281 |
| IUPAC Name | 2-chloro-1-phenylpropan-1-one |
| SMILES | CC(Cl)C(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 6084-17-9 | |
| 168.62 | |
| AXCPQHPNAZONTH-UHFFFAOYNA-N | |
| 221281 | |
| CC(Cl)C(=O)C1=CC=CC=C1 |
| C9H9ClO | |
| MFCD03844782 | |
| 2-Chloro-1-phenyl-1-propanone | |
| 2-chloro-1-phenylpropan-1-one |
Specifications
| 6084-17-9 | |
| C9H9ClO | |
| 5 g | |
| AXCPQHPNAZONTH-UHFFFAOYNA-N | |
| 2-chloro-1-phenylpropan-1-one | |
| 221281 | |
| ≥95.0% (GC) | |
| 2-Chloropropiophenone |
| Yellow | |
| MFCD03844782 | |
| 2-Chloro-1-phenyl-1-propanone | |
| CC(Cl)C(=O)C1=CC=CC=C1 | |
| 168.62 | |
| 168.62 | |
| Liquid |
Safety and Handling
TSCA : No