missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloropropiophenone 96.0+%, TCI America™

Catalog No. c33231g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33231G Supplier TCI America Supplier No. C33231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6323-18-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD07787256
InChI Key BTSCBJDORATYKJ-UHFFFAOYSA-N
Synonym 2-Chlorophenyl Ethyl Ketone
PubChem CID 95743
IUPAC Name 1-(2-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 116°C
Quantity 1 g
Formula Weight 168.62
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Chloropropiophenone